Learn More
BUCHI FlashPure ID Diol Available on GSA/VA Contract for Federal Government customers only.
FlashPure ID cartridges are degined for Reveleris™ chromatography systems. Integrated RFID technology automatically transfers cartridge settings and data to the flash system. Diol functionalized silica provides an alternate selectivity for a variety of polarity types and is ideal for lipids.
$456.00 - $3073.00
Specifications
For Use With (Application) Alternate Selectivity for a Variety of Polar Types, Ideal for Lipids
For Use With (Equipment) Reveleris Flash System
Material Polypropylene
Type Diol
Column Type Diol
View More Specs
Products 3
Catalog Number Mfr. No. Size (Metric) Tube Rack Includes Price Quantity & Availability  
Catalog Number Mfr. No. Size (Metric) Tube Rack Includes Price Quantity & Availability  
05-410-116 Available on GSA/VA Contract for Federal Government customers only. View Documents BUCHI
140000150
220 g 1 Pc. Pack for $2,639.00
Add to cart
05-410-001 Available on GSA/VA Contract for Federal Government customers only. View Documents BUCHI
140000018
24 g 1 Pc. Pack for $456.00
Add to cart
05-410-117 Available on GSA/VA Contract for Federal Government customers only. View Documents BUCHI
140000151
330 g 1 Pc. Pack for $3,073.00
Add to cart
Description
Description
  • RFID for optimal convenience on BUCHI Reveleris™ systems
  • 40 µm irregular particle
  • Selection of cartridge sizes from 4g-330g
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Alternate Selectivity for a Variety of Polar Types, Ideal for Lipids
Polypropylene
Diol
Reveleris Flash System
Diol