Learn More
BUCHI FlashPure ID HP Si
FlashPure ID cartridges are degined for Reveleris™ chromatography systems. Integrated RFID technology automatically transfers cartridge settings and data to the flash system. Silica HP provides higer performance for polar compounds using organic solvents.
Manufacturer:  BUCHI 145170208
Catalog No. 05-410-122
Description
Description
  • RFID for optimal convenience on BUCHI Reveleris™ systems
  • 20 µm irregular particle
  • Selection of cartridge sizes from 4g-330g
Specifications
Specifications
Polypropylene
4 Pc.
330 g
Alternate Selectivity for a Variety of Polar Types, Ideal for Lipids
High Performance Silica
16 to 24 μm
Silica
Reveleris Flash System