Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BUCHI FlashPure ID Silica-Coated TLC Plates with Fluorescent Indicator

Glass or aluminum backed plates coated with FlashPure ID Silica with fluorescent indicator, UV 254 nm.
Supplier: BUCHI 148621250
Description
- Glass or aluminum backed plates
- Coated with FlashPure ID Silica
- For normal phase applications
Specifications
Glass | |
254 nm | |
Normal Phase | |
0.74 to 0.76 mL/g | |
20 x 20 cm |
5 to 17 μm | |
Yes | |
Silica | |
FlashPure |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title