Learn More
BUCHI FlashPure ID Silica Flash Cartridges PROMO
Excels in conventional applications
Manufacturer:  BUCHI 145165730PM
 View more versions of this product
Catalog No. 05-404-837PM
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
High performance
  • High resolution of separation due to high-quality silica
  • Increased fraction purity due to a better separation
  • Faster evaporation of smaller fraction volumes
  • Reproducible performance from batch to batch
Lower purification costs
  • Increased sample throughput due to high sample loading capacity
  • Solvent saving: required purification yield is achieved faster
  • /ul>
Specifications
Specifications
24 g
Useful for Conventional Applications; Polar Compounds
15 Pc.
Silica