Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUCHI FlashPure Select Amino
SDP

FlashPure Select Amino cartridges are designed for ultimate and unmatched performance. Amino functionalized silica provides better peak shapes with polar and basic compounds and is ideal for carbohydrates.

Manufacturer:  BUCHI 140000133

 View more versions of this product

Catalog No. 05-410-099

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • 30 μm spherical particles
  • Functionalized silica cartridges can be reused multiple times
  • Large selection of cartridge sizes from 4g-330g
  • Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen.
Specifications

Specifications

Polypropylene
2 Pc.
80 g
Better Peak Shapes with Polar and Basic Compounds, Ideal for Carbohydrates
Amino
20 to 35 μm
Amino
Any Flash System
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit