Learn More
BUCHI FlashPure Select Amino
FlashPure Select Amino cartridges are designed for ultimate and unmatched performance. Amino functionalized silica provides better peak shapes with polar and basic compounds and is ideal for carbohydrates.
Specifications
For Use With (Application) Better Peak Shapes with Polar and Basic Compounds, Ideal for Carbohydrates
For Use With (Equipment) Any Flash System
Material Polypropylene
Type Amino
Column Type Amino
View More Specs
Products 8
Catalog Number Mfr. No. Size (Metric) Tube Rack Includes Price Quantity & Availability  
Catalog Number Mfr. No. Size (Metric) Tube Rack Includes Price Quantity & Availability  
05-410-100
View Documents
BUCHI
140000134
120 g 2 Pc. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
05-410-096
View Documents
BUCHI
140000130
12 g 5 Pc. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
05-410-097
View Documents
BUCHI
140000131
20 g 2 Pc. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
05-410-101
View Documents
BUCHI
140000135
220 g 1 Pc. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
05-410-102
View Documents
BUCHI
140000136
330 g 1 Pc. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
05-410-098
View Documents
BUCHI
140000132
40 g 2 Pc. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
05-410-095
View Documents
BUCHI
140000129
4 g 5 Pc. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
05-410-099
View Documents
BUCHI
140000133
80 g 2 Pc. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • 30 μm spherical particles
  • Functionalized silica cartridges can be reused multiple times
  • Large selection of cartridge sizes from 4g-330g
  • Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen.
Specifications
Specifications
Better Peak Shapes with Polar and Basic Compounds, Ideal for Carbohydrates
Polypropylene
Amino
Any Flash System
Amino