Learn More
BUCHI FlashPure Select Diol
FlashPure Select Diol cartridges are designed for ultimate and unmatched performance. Diol functionalized silica provides an alternate selectivity for a variety of polarity types and is ideal for lipids.
Manufacturer:  BUCHI 140000142
 View more versions of this product
Catalog No. 05-410-108
Add to cart
Description
Description
  • 30 µm spherical particles
  • Functionalized silica cartridges can be reused multiple times
  • Large selection of cartridge sizes from 4g-330g
  • Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen.
Specifications
Specifications
Diol
Any Flash System
Polypropylene
120 g
Alternate Selectivity for a Variety of Polar Types, Ideal for Lipids
2 Pc.
20 to 35 μm
Diol