Learn More
BUCHI FlashPure Select Diol
FlashPure Select Diol cartridges are designed for ultimate and unmatched performance. Diol functionalized silica provides an alternate selectivity for a variety of polarity types and is ideal for lipids.
Specifications
For Use With (Application) Alternate Selectivity for a Variety of Polar Types, Ideal for Lipids
For Use With (Equipment) Any Flash System
Material Polypropylene
Type Diol
Column Type Diol
View More Specs
Products 1
Catalog Number Mfr. No. Size (Metric) Tube Rack Includes Price Quantity & Availability  
Catalog Number Mfr. No. Size (Metric) Tube Rack Includes Price Quantity & Availability  
05-410-103
View Documents
BUCHI
140000137
4 g 5 Pc. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • 30 μm spherical particles
  • Functionalized silica cartridges can be reused multiple times
  • Large selection of cartridge sizes from 4g-330g
  • Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen.
Specifications
Specifications
Alternate Selectivity for a Variety of Polar Types, Ideal for Lipids
Polypropylene
Diol
Any Flash System
Diol