Fisherbrand™ Flask Covers Available on GSA/VA Contract for Federal Government customers only.
Cut down evaporation
 View more versions of this product
Catalog No. 10-042A
  Check Availability
Add to Cart
    Specifications
    Product Type Cover, Flask
    For Use With (Equipment) Use with flask that takes Rubber stopper sizes, used when solutions are being boiled to cause condensation and refluxing
    Stopper Number 6
    Closure Material Flint Glass
  • Description & Specifications
Related Products
Product Content Feedback