Learn More
Fisherbrand™ Flask Covers
SDP
Cut down on evaporation.
 View more versions of this product
Catalog No. 10-042B
Add to Cart
Description
Description
  • Flint glass
  • Designed by Professor F.E. Tuttle
  • Place in necks of flasks while solutions are being boiled to cause condensation and refluxing
  • Use with any flask that takes rubber stopper sizes listed in ordering table
Specifications
Specifications
Flint Glass
Use with flask that takes Rubber stopper sizes, used when solutions are being boiled to cause condensation and refluxing
7 - 8
Glass
Cover, Flask
Product Suggestions
Product Suggestions