DFS Item Hach Company FLASK DIGESDAHL W/STOPPER PKG
Manufacturer: Hach Company 2312542
Flask, Flat-Bottom, Calibrated Individually with a 100mL (to contain) Mark
Catalog No. NC9593729
 N/A
N/A
Please call customer service for current pricing. 1-800-766-7000 
Related Products
Product Content Feedback