Learn More
Wilmad Labglass FLASK KJELDAHL 500ML 29/42
Manufacturer:  Wilmad Labglass LG7920112
Encompass_Preferred
Kjeldahl Flask, Long Neck, 500mL capacity. Joint Size: [ST] 29/42 outer. Kjeldahl shaped flask w/ a standard taper outer joint.
Catalog No. 50-803-077
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.