Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synthware Reaction Tubes with Glass Stopcock
Cylindrical shaped tube with standard taper outer joint
Manufacturer: Synthware F894100
Description
- Side arm with 2mm glass stopcock
- Stopcock supplied with constant pressure retention device
Specifications
Reaction Tube with Glass Stopcock | |
100 mL | |
Glass | |
24/40 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title