Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversified Spaces™ Flat File Storage
SCI_ED

Protect and store your art work, drawings or biological collections. The Flat File Storage can sit on a counter or be stacked up to three units.

Supplier:  Diversified Spaces™ P7202BM30N

 View more versions of this product

Catalog No. S47345


Add to Cart

Description

Description

  • 4 in. toe kick not included P/N #FFSB-3624
  • 40-1/2 in.W x 27 in.D x 17 in.H
  • Diversified Spaces™ is SEFA compliant and MAS Certified Green
  • Five drawers with inside dimensions measure 37 in.W x 24-13/16 in.D x 1-1/2 in.H
  • Full extension drawer glides
  • Limited Lifetime Warranty
  • Made in the USA
  • The unit is finished with a chemical resistant, earth-friendly UV finish
  • Top allows for the Flat File System Top P/N #FFST to be added
  • Unit is designed to sit on a counter or on the Flat File System Base P/N #FFSB

Warranty and Services

Lifetime Limited Warranty

Specifications

Specifications

File Storage
27 in.
17in.
37 in.
5
40-1/2 in.
24-13/16 in.
Natural
Papers up to 24 x 36in.
1.5 in.
Flat
Maple
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit