Learn More
Flat Glass Plates Products for Science Education
Explore thin film interference and develop a way to measure the thickness of sample materials enclosed.
Manufacturer:  Science FirstSupplier Diversity Partner 15040
Catalog No. S52059
Add to cart
Description
Description
  • Plates are sandwiched to include the sample on one side, forming an air wedge
  • Requires a monochromatic light source
Specifications
Specifications
Explore thin film interference and develop a way to measure the thickness of sample materials enclosed
Glass
Flat Plate