Learn More
Simport™ Scientific Flat Top Caps for Microcentrifuge Tubes
Simport™ Scientific Flat Top Caps for Microcentrifuge Tubes with silicone washer seal is molded with deep internal lip that fits snugly against interior wall of tube thus reducing chances of sample contamination.
Manufacturer:  Simport™ Scientific T340OOSFT
Catalog No. 23-045-257
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Made of polypropylene
  • Flat top with silicone washer seal
  • Autoclavable
  • Will withstand temperatures from -196°C to +121°C
  • Certified RNase, DNase, DNA and Pyrogen free
  • Orange
Specifications
Specifications
Micrewtube Microcentrifuge Tube
Autoclavable
Orange
No
Flat Top with Washer Seal
Polypropylene
Silicone
Non-sterile