Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Flavanone, 98+%

Click to view available options
Quantity:
10g
50g
487-26-3
C15H12O2
224.259
MFCD00006841
ZONYXWQDUYMKFB-UHFFFAOYSA-N
flavanone,2-phenylchroman-4-one,2,3-dihydroflavone,2-phenyl-4-chromanone,4-flavanone,2,3-dihydro-2-phenyl-4h-1-benzopyran-4-one,4h-1-benzopyran-4-one, 2,3-dihydro-2-phenyl,2-phenyl-2,3-dihydro-4h-chromen-4-one,2,3-dihydro-2-phenyl-4h-benzopyran-4-one,2-phenyl-3,4-dihydro-2h-1-benzopyran-4-one
10251
CHEBI:5070
2-phenyl-2,3-dihydrochromen-4-one
C1C(OC2=CC=CC=C2C1=O)C3=CC=CC=C3
487-26-3
C15H12O2
224.259
MFCD00006841
ZONYXWQDUYMKFB-UHFFFAOYSA-N
flavanone,2-phenylchroman-4-one,2,3-dihydroflavone,2-phenyl-4-chromanone,4-flavanone,2,3-dihydro-2-phenyl-4h-1-benzopyran-4-one,4h-1-benzopyran-4-one, 2,3-dihydro-2-phenyl,2-phenyl-2,3-dihydro-4h-chromen-4-one,2,3-dihydro-2-phenyl-4h-benzopyran-4-one,2-phenyl-3,4-dihydro-2h-1-benzopyran-4-one
10251
CHEBI:5070
2-phenyl-2,3-dihydrochromen-4-one
C1C(OC2=CC=CC=C2C1=O)C3=CC=CC=C3
Quantity 10g
Melting Point 75°C to 79°C
Beilstein 183227
Formula Weight 224.26
Percent Purity ≥98%
Chemical Name or Material Flavanone

RTECSNumber : LK6906500

TSCA : TSCA