Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flavin Adenine Dinucleotide Disodium Salt Hydrate, ≥95% (dry basis), Affymetrix/USB™

Click to view available options
Quantity:
500mg
1g
Packaging:
Plastic bottle
84366-81-4
C27H33N9Na2O16P2
847.535
MFCD00151217
GXTPHHZYFMAGLX-UJXBNFGUSA-L
flavin adenine dinucleotide disodium,unii-67u7uhj04c,flavin adenine dinucleotide disodium salt hydrate,riboflavin 5'-adenosine diphosphate disodium salt,flavin adenine dinucleotide disodium salt,fad-na2,flavine adenine dinucleotide,fad sodium,flavin adenine dinucelotide,fad tn
131675332
disodium;[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4S)-5-(7,8-dimethyl-4-oxido-2-oxobenzo[g]pteridin-10-yl)-2,3,4-trihydroxypentyl] phosphate;hydrate
CC1=CC2=C(C=C1C)N(C3=NC(=O)N=C(C3=N2)[O-])CC(C(C(COP(=O)([O-])OP(=O)(O)OCC4C(C(C(O4)N5C=NC6=C5N=CN=C6N)O)O)O)O)O.O.[Na+].[Na+]
84366-81-4
C27H33N9Na2O16P2
847.535
MFCD00151217
GXTPHHZYFMAGLX-UJXBNFGUSA-L
flavin adenine dinucleotide disodium,unii-67u7uhj04c,flavin adenine dinucleotide disodium salt hydrate,riboflavin 5'-adenosine diphosphate disodium salt,flavin adenine dinucleotide disodium salt,fad-na2,flavine adenine dinucleotide,fad sodium,flavin adenine dinucelotide,fad tn
131675332
disodium;[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4S)-5-(7,8-dimethyl-4-oxido-2-oxobenzo[g]pteridin-10-yl)-2,3,4-trihydroxypentyl] phosphate;hydrate
CC1=CC2=C(C=C1C)N(C3=NC(=O)N=C(C3=N2)[O-])CC(C(C(COP(=O)([O-])OP(=O)(O)OCC4C(C(C(O4)N5C=NC6=C5N=CN=C6N)O)O)O)O)O.O.[Na+].[Na+]
Formula Weight 829.51 (anhydrous)
Physical Form Crystalline Powder
Percent Purity ≥93%
Packaging Plastic bottle
Quantity 500mg
Chemical Name or Material Flavin Adenine Dinucleotide Disodium Salt Hydrate
Assay Dry basis