Learn More
Flavivirus group antigen Mouse anti-Virus, Alexa Fluor™ 750, Clone: D1-4G2-4-15 (4G2), Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP252709AF750
Catalog No. NP22709A70
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Flavivirus group antigen Monoclonal antibody specifically detects Flavivirus group antigen in Virus samples. It is validated for Western Blot,Flow Cytometry,ELISA,Immunohistochemistry,Immunohistochemistry (Paraffin),Immunofluorescence.
Specifications
Specifications
Flavivirus group antigen
D1-4G2-4-15 (4G2)
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence
Mouse
IgG2a Kappa
Store at 4°C in the dark.
Monoclonal
Virus
ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
Dengue Virus type 2 antigens.
0.1 mL
Primary
Recognises flavivirus group specific antigens (Dengue virus, West Nile Virus, Japanese Encephalitis, Yellow Fever Virus, Zika virus etc). It binds to the fusion loop at the extremity of domain II of protein E.