Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flavivirus group antigen Rabbit anti-Virus, Alexa Fluor™ 350, Clone: D1-4G2-4-15 (4G2), Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP252666AF350

Catalog No. NP22666A30


Add to cart

Description

Description

Flavivirus group antigen Monoclonal antibody specifically detects Flavivirus group antigen in Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence.
Specifications

Specifications

Flavivirus group antigen
D1-4G2-4-15 (4G2)
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence
Conjugated, Purified
Dengue Virus type 2 antigens.
Protein A or G purified
Store at 4°C in the dark.
Monoclonal
Virus
ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
Rabbit
IgG Kappa
0.1 mL
Primary
Recognises flavivirus group specific antigens (Dengue virus, West Nile Virus, Japanese Encephalitis, Yellow Fever Virus, Zika virus etc). It binds to the fusion loop at the extremity of domain II of protein E.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit