Learn More
Flavivirusgroupantigen Rabbit anti-Viral, Janelia Fluor 549, Clone: D1-4G2-4-15 (4G2), Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP252666JF549
Catalog No. NBP52666549
Add to cart
Description
Description
Flavivirusgroupantigen Monoclonal antibody specifically detects Flavivirusgroupantigen in Viral samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
Specifications
Flavivirusgroupantigen
D1-4G2-4-15 (4G2)
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence
Purified
Dengue Virus type 2 antigens.
Protein A or G purified
Store at 4°C in the dark.
Monoclonal
Viral, Virus
Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
Rabbit
IgG Kappa
0.1mL
Primary
Recognises flavivirus group specific antigens (Dengue virus, West Nile Virus, Japanese Encephalitis, Yellow Fever Virus, Zika virus etc). It binds to the fusion loop at the extremity of domain II of protein E.