Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flavone, 99+%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
525-82-6
C15H10O2
222.24
VHBFFQKBGNRLFZ-UHFFFAOYSA-N
flavone,2-phenylchromone,2-phenyl-4h-chromen-4-one,2-phenyl-4-chromone,2-phenyl-4-benzopyron,2-phenyl-4h-1-benzopyran-4-one,4h-1-benzopyran-4-one, 2-phenyl,asmacoril,chromocor,cromaril
10680
CHEBI:42491
2-phenylchromen-4-one
C1=CC=C(C=C1)C2=CC(=O)C3=CC=CC=C3O2
525-82-6
C15H10O2
222.24
VHBFFQKBGNRLFZ-UHFFFAOYSA-N
flavone,2-phenylchromone,2-phenyl-4h-chromen-4-one,2-phenyl-4-chromone,2-phenyl-4-benzopyron,2-phenyl-4h-1-benzopyran-4-one,4h-1-benzopyran-4-one, 2-phenyl,asmacoril,chromocor,cromaril
10680
CHEBI:42491
2-phenylchromen-4-one
C1=CC=C(C=C1)C2=CC(=O)C3=CC=CC=C3O2
Infrared Spectrum Authentic
Melting Point 95.0°C to 98.0°C
Packaging Glass bottle
Quantity 5g
Color White
Assay Percent Range 99+%
Beilstein 17, 373
Merck Index 15, 4124
Solubility Information Solubility in water: practically insoluble in water. Other solubilities: soluble in most common organic solvents
Formula Weight 222.24
Physical Form Crystalline Powder
Percent Purity ≥99%
Chemical Name or Material Flavone
Show More Show Less