Learn More
Selleck Chemical LLCSupplier Diversity Partner Flavopiridol HCl DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S267910MM/1ML
Flavopiridol HCl competes with ATP to inhibit CDKs including CDK1, CDK2, CDK4 and CDK6 with IC50 of ~ 40 nM in cell-free assays. It is 7.5-fold more selective for CDK1/2/4/6 than CDK7. Flavopiridol is initially found to inhibit EGFR and PKA. Phase 1/2.
Catalog No. 50-194-7960