Learn More
Pyramex™ Flex-Lyte™ Safety Glasses
Pyramex™ Flex-Lyte™ Safety Glasses with ANSI Z87 safety compliance
$2.15 - $2.55
Specifications
Certifications/Compliance ANSI Z87
For Use With (Application) Eye Safety
Frame Material Polycarbonate
Lens Material Polycarbonate
Temple Style Slim Co-Injected
View More Specs
Products 6
Catalog Number Mfr. No. Frame Color Lens Tint Price Quantity & Availability  
Catalog Number Mfr. No. Frame Color Lens Tint Price Quantity & Availability  
19-555-683
View Documents
Pyramex™
SBG10510D
Black, Gray Clear
Each for $2.15
Case of 12 EA for $25.80
Add to cart
19-555-685
View Documents
Pyramex™
SBG10520D
Black, Gray Gray
Each for $2.40
Case of 12 EA for $28.80
Add to cart
19-555-687
View Documents
Pyramex™
SBG10580D
Black, Gray Indoor/Outdoor, Mirror
Each for $2.55
Case of 12 EA for $30.60
Add to cart
19-555-696
View Documents
Pyramex™
SBL10510D
Black, Lime Clear
Each for $2.15
Case of 12 EA for $25.80
Add to cart
19-555-698
View Documents
Pyramex™
SBL10520D
Black, Lime Gray
Each for $2.40
Case of 12 EA for $28.80
Add to cart
19-555-700
View Documents
Pyramex™
SBL10580D
Black, Lime Indoor/Outdoor, Mirror
Each for $2.55
Case of 12 EA for $30.60
Add to cart
Description
Description
  • Soft nose piece for snug fit
  • Semi-frame & slim co-material temple design for lightweight spectacle - 22 gm
  • Offers a clear, panoramic view without obstruction
  • 9 base wraparound lens provides full front side protection
  • Scratch-resistant, polycarbonate lens
  • Lens Technology:
    • Visible Light Transmitted - 96%
    • UVA, UVB Blocked - 100%
    • Blue Light Blocked - 9%
    • IR Blocked - 18%
Specifications
Specifications
ANSI Z87
Polycarbonate
Slim Co-Injected
Anti-fog
Eye Safety
Polycarbonate
1