Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flexible Collodion, USP, Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
2.5 L
4 L
200 L
Packaging:
Amber Glass Bottle
Drum
9004-70-0,64-17-5
C24H36N8O38
1044.57
OWWGYSXLINVAES-UHFFFAOYNA-N
2,3-bis(nitrooxy)-6-[(nitrooxy)methyl]-5-{[3,4,5-tris(nitrooxy)-6-[(nitrooxy)methyl]oxan-2-yl]oxy}oxan-4-yl nitrate; 2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O.[O-][N+](=O)OCC1OC(O[N+]([O-])=O)C(O[N+]([O-])=O)C(O[N+]([O-])=O)C1OC1OC(CO[N+]([O-])=O)C(O[N+]([O-])=O)C(O[N+]([O-])=O)C1O[N+]([O-])=O
9004-70-0,64-17-5
C24H36N8O38
1044.57
OWWGYSXLINVAES-UHFFFAOYNA-N
2,3-bis(nitrooxy)-6-[(nitrooxy)methyl]-5-{[3,4,5-tris(nitrooxy)-6-[(nitrooxy)methyl]oxan-2-yl]oxy}oxan-4-yl nitrate; 2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O.[O-][N+](=O)OCC1OC(O[N+]([O-])=O)C(O[N+]([O-])=O)C(O[N+]([O-])=O)C1OC1OC(CO[N+]([O-])=O)C(O[N+]([O-])=O)C(O[N+]([O-])=O)C1O[N+]([O-])=O

CO125

Spectrum™ Flexible Collodion, USP is used as a surgical dressing and forms a flexible nitrocellulose membrane. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Drum
Quantity 200 L
CAS Max % 0.001,0.999
UN Number UN2059