Learn More
Sartorius™ Flexible Sample Holder, for Cubis™ Balances Available on GSA/VA Contract for Federal Government customers only.
For use with Cubis Balances. Q-Grip is a flexible and adaptable “one-size-fits-all” holder for bottles, test tubes, reaction containers or filters (up to 120mm) for all Cubis semi-microbalances and analytical balances.
Manufacturer:  Sartorius™ YFH01MS
Catalog No. 14-557-554
Add to cart
Description
Description
Includes:
  • Plate
  • Filter supports (2)
  • Flask supports (2)
  • Pan support
  • Clamping screw and nut
Replaces the original weighing pan and is only for CUBIS Balances with 0.1 and 0.01mg readability.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Weighting Flask Holder
Containers and filters of Cubis™ semi-microb
Documents
Promotions
Promotions
  • Sartorius , Fisherbrand , Fisher Scientific
    Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.
    06-30-2021