Learn More
Ready America™ QuakeHOLD!™ Flex Mount Black Fastener
SDP
Ready America™ QuakeHOLD!™ Flex Mount Black Fastener is suitable for bracing flat surface equipment by attaching to your unit with the opposing end secured to the flexmount bar.
Supplier:  Ready America™ RDFMT42
Catalog No. 11-900-052
Add to Cart
Description
Description
  • Secures flat surface equipment
  • Used in conjunction with Flex Mount Bar
  • Ideal for IBC Compliance
  • Features 3M™ VHB adhesive
  • 100 lb weight capacity
Product Suggestions
Product Suggestions