Learn More
Sartorius Flexsafe RM 50L basic (5 pcs)
Supplier:  Sartorius DFB050L
Encompass
Flexsafe RM bag is a single-use bioreactor bag and a member of our bioprocessing bag Flexsafe family with outstanding film quality. Benefit from the same polyethylene film material across all your cell culture steps. The bags are mainly used for an easy and quick seed train and scale up to our BIOSTAT STR single-use stirred tank bioreactor equipped with Flexsafe STR bags. Flexsafe RM bags fit on rocking motion bioreactors from several manufacturers.
Catalog No. NC1540093