Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wells Lamont™ Flextech™ Polyurethane Coated Synthetic Knit Gloves
Click to view available options
Size:
2X-Large
Large
Medium
Small
X-Large
X-Small
Description
- 13 gauge, synthetic knit shell increases dexterity and flexibility
- Polyurethane coating on the palm enhances grip capabilities
- Open, uncoated back circulates air to allow hands to breathe
- Non-slip surface allows workers to grip objects with confidence
- Silicone-free
Specifications
ANSI Abrasion Rating | ANSI 4 |
Construction | Seamless Knit |
Coating Material | Polyurethane |
Color | White |
Hand | Specific |
Latex Free | No |
Material | Nylon |
Product Type | Seamless Knit Gloves |
Size | Medium |
Glove Cuff Color | Yellow |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction