Tocris™ FLI 06

Inhibitor of Notch signaling

Manufacturer: Tocris™ 5172/50

 View more versions of this product

Catalog No. 51-725-0

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Molecular Weight 438.52
    Quantity 50mg
    Inhibitors FLI 06
    Purity >99
    Product Type FLI 06

    Inhibitor of Notch signaling (EC50 = 2.3µM). Disrupts Notch trafficking and processing. Reduces amyloid β secretion. Changes the maturation and abolishes shredding of APP. Inhibits ER exporting and converts tubular ER to sheets.

    • Chemical Name: Cyclohexyl 1,4,5,6,7,8-hexahydro-2,7,7-trimethyl-4-(4-nitrophenyl)-5-oxo-3-quinolinecarboxylate
    • Formula: C25H30N2O5
    • CAS Number: 313967-18-9
    • Solubility: Soluble to 100mM in DMSO and to 20mM in ethanol
    • Purity: >99

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit