Learn More
Tocris Bioscience™ FLI 06
Inhibitor of Notch signaling; disrupts Notch trafficking and processing
Manufacturer:  Tocris Bioscience™ 5172/10
 View more versions of this product
Catalog No. 51-721-0
Add to cart
This item is not returnable. View return policy
Description
Description
Inhibitor of Notch signaling (EC50 = 2.3 μM). Disrupts Notch trafficking and processing. Reduces amyloid β secretion. Changes the maturation and abolishes shredding of APP. Inhibits ER exporting and converts tubular ER to sheets.
Specifications
Specifications
FLI 06
>99
10mg
FLI 06
438.52