Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Fli-1, Rabbit Polyclonal Antibody

Obtain accurate, reproducible results in western blotting experiments with Thermo Scientific™ Fli-1, Rabbit Polyclonal Antibody.

Specifications

Antigen Fli-1
Concentration 200µg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
RB9295P1
View Documents
Thermo Scientific™
RB9295P1
Purified Ab with BSA and Azide 500μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Obtain accurate, reproducible results in western blotting experiments with Thermo Scientific™ Fli-1, Rabbit Polyclonal Antibody.

Ets-1 is the prototype member of a family of genes identified on the basis of homology to the v-Ets oncogene isolated from the E26 erythroblastosis virus. This family of genes currently includes Ets-1, Ets-2, Erg-1, Erg-2, Elk, E74, Fli-1, PU.1 and PEA3. Members of the Ets gene family exhibit varied patterns of tissue expression and share a highly conserved carboxy terminal domain containing a sequence related to the SV40 large T antigen nuclear localizartion signal sequence.

Host Species: Rabbit

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: C-terminal

Immunogen: Synthetic peptide derived from C-terminus of human Fli-1

Molecular Weight: ∽50 kDa

Positive Control: Ewing's sarcoma, Angiosarcoma, Jurkat cells

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Fli-1
Western Blot
Unconjugated
Rabbit
Transcription Regulation
Purified Ab with BSA and Azide
200µg/mL
Polyclonal
Concentrated
RUO
Primary
Synthetic peptide derived from C-terminus of human Fli-1
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit