Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLI1 Mouse anti-Human, Clone: FLI1/1312, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14785

Catalog No. 89-034-868


Add to cart

Description

Description

Mouse monoclonal antibody raised against partial recombinant human FLI1.

  • High specificity
  • High reactivity
Specifications

Specifications

FLI1
FLI1/1312
Mouse monoclonal antibody raised against partial recombinant human FLI1.
In 10 mM PBS.
Liquid
FLI1
Recombinant protein corresponding to amino acids 107-261 of human FLI1.
Protein A purification
RUO
Primary
2313
Flow Cytometry, Immunofluorescence, Western Blot
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (1-2 ug/mL) Western Blotting (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Q01543
EWSR2|SIC-1
Mouse
IgG
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)