Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FLI1 Mouse anti-Human, Clone: FLI1/3183, Novus Biologicals™
Mouse Monoclonal Antibody
$265.00
Specifications
Antigen | FLI1 |
---|---|
Clone | FLI1/3183 |
Concentration | 0.2 mg/ml |
Dilution | ELISA, Protein Array |
Applications | ELISA, Peptide Array |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB122330
|
Novus Biologicals
NBP30742820UG |
20 μg |
Each of 1 for $265.00
|
|
|||||
Description
FLI1 Monoclonal specifically detects FLI1 in Human samples. It is validated for ELISA, Protein Array.Specifications
FLI1 | |
0.2 mg/ml | |
ELISA, Peptide Array | |
Unconjugated | |
Mouse | |
Cancer, Tumor Suppressors | |
10 mM PBS with 0.05% BSA | |
2313 | |
IgG1 κ | |
Protein A purified |
FLI1/3183 | |
ELISA, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EWSR2, Friend leukemia integration 1 transcription factor, Friend leukemia virus integration 1, Proto-oncogene Fli-1, SIC-1, Transcription factor ERGB | |
Recombinant fragment (aa 107-261) of human FLI1 protein with hexa-histidine tag | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title