Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLI1 Mouse anti-Human, PE/Atto594, Clone: FLI1/1312, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254423PEATT594

Catalog No. N254423P594


Add to cart

Description

Description

FLI1 Monoclonal antibody specifically detects FLI1 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

FLI1
FLI1/1312
Flow Cytometry
Conjugated, Purified
FLI1
Recombinant fragment (aa 107-261) of human FLI1 protein (exact sequence is proprietary)
Protein A purified
Cancer, Tumor Suppressors
Primary
2313.0
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
EWSR2, Friend leukemia integration 1 transcription factor, Friend leukemia virus integration 1, Proto-oncogene Fli-1, SIC-1, Transcription factor ERGB
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit