Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLIP Mouse anti-Human, Mouse, DyLight 488, Clone: 7A3D12, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237359G

Catalog No. NBP237359G

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

FLIP
7A3D12
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Purified
CFLAR
Purified recombinant fragment of human CFLAR (AA: 100-251) expressed in E. Coli.
Protein A or G purified
Apoptosis, Cancer, Cell Biology, Inhibitors Activators and Regulators, Signal Transduction, Tumor Suppressors
Primary
8837
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
CASHFADD-like antiapoptotic molecule 1, CASP8 and FADD-like apoptosis regulator, CASP8AP1, Caspase homolog, Caspase-eight-related protein, Caspase-like apoptosis regulatory protein, caspase-related inducer of apoptosis, CASPER, CasperFLAME1, Cellular FLICE-like inhibitory protein, c-FLIPR, c-FLIPS, CLARPMACH-related inducer of toxicity, FADD-like apoptotic molecule, FLAME, FLAME-1, I-FLICEMRITc-FLIPFLIPc-FLIPL, Inhibitor of FLICE, Usurpin, usurpin beta
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit