Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ FlipStrip™ Microtube Racks with Lid

Hold 0.2mL strips and 1.5mL tubes on one side and and 0.5mL tubes on the other side of this microtube rack with lid, which is made from 100% recycled plastic.

Supplier:  Thermo Scientific™ 8638

 View more versions of this product

Catalog No. 21-402-32

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

Hold 0.2mL strips and 1.5mL tubes on one side of the Thermo Scientific™ FlipStrip Microtube Rack with Lid, and 0.5mL tubes on the other. The rack is made from 100% recycled plastic.

  • Holds 0.2mL strips and 1.5mL tubes on one side, 0.5mL tubes on other
  • Stable, low-profile design with an alphanumeric grid for easy tube identification
  • Lid protects samples and allows racks to be stacked
  • Several colors available
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Flipstrip Rack
Tubes (0.5 mL), Tubes (1.5 mL), Tubes (2 mL)
Yes
Blue
Recycled Plastic
12 Racks
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit