Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FLJ10808 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP224534AF350
Description
FLJ10808 Polyclonal specifically detects FLJ10808 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| FLJ10808 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| E1-L2, FLJ10808, FLJ23367, Monocyte protein 4, MOP4, MOP-4, UBA6, ubiquitin-activating enzyme E1, UBE1L2, Ubiquitin-activating enzyme 6, ubiquitin-activating enzyme E1-like 2, Ubiquitin-activating enzyme E1-like protein 2, ubiquitin-like modifier activating enzyme 6, ubiquitin-like modifier-activating enzyme 6 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| UBA6 | |
| A portion of amino acids 775-825 of human UBE1L2 was used as the immunogen for this antibody. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction