Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLJ10808 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen FLJ10808
Applications Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB092539
View Documents
Novus Biologicals
NBP303402100UL
100 μg Each for $320.71
Add to cart
 
NB092540
View Documents
Novus Biologicals
NBP30340220UL
20 μg Each for $138.37
Add to cart
 
Description

Description

FLJ10808 Polyclonal antibody specifically detects FLJ10808 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Specifications

Specifications

FLJ10808
Unconjugated
Rabbit
Human, Mouse, Rat
E1-L2, FLJ10808, FLJ23367, Monocyte protein 4, MOP4, MOP-4, UBA6, ubiquitin-activating enzyme E1, UBE1L2, Ubiquitin-activating enzyme 6, ubiquitin-activating enzyme E1-like 2, Ubiquitin-activating enzyme E1-like protein 2, ubiquitin-like modifier activating enzyme 6, ubiquitin-like modifier-activating enzyme 6
Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human FLJ10808 (NP_060697.4).
Polyclonal
Affinity purified
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
55236
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit