Learn More
Aviva Systems Biology Corp FLJ25439 Antibody - N-terminal region (ARP52760_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP52760_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FLJ25439. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FLJ25439. Alternate names: MC25-1, FLJ25439ApplicationWB. Species reactivity: Cow, Horse, Human, Rabbit
Catalog No. 50-014-60824