Learn More
Aviva Systems Biology Corp FLJ40504 ANTIBODY 50UG/50UL
Supplier:  Aviva Systems Biology Corp ARP55671_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FLJ40504. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FLJ40504. Alternate names: MGC138231, MGC138233, FLJ40504ApplicationWB. Species reactivity: Human
Catalog No. 50-014-61573
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.