Learn More
FLJ40504 Mouse anti-Human, Clone: OTI11D8, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF810104
Catalog No. 50-166-9890
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
FLJ40504 encodes the type I intermediate filament chain keratin 18. Keratin 18, together with its filament partner keratin 8, are perhaps the most commonly found members of the intermediate filament gene family. They are expressed in single layer epithelial tissues of the body. Mutations in this gene have been linked to cryptogenic cirrhosis. Two transcript variants encoding the same protein have been found for this gene.
Specifications
Specifications
FLJ40504
Monoclonal
Unconjugated
KRT18P55
keratin 18 pseudogene 55
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
284085
Western Blot
OTI11D8
PBS with 8% trehalose and no preservative; pH 7.3
Lyophilized
KRT18P55
Full length human recombit protein of human FLJ40504 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human