Learn More
FLK-1 Rat anti-Mouse, BV421, Clone: Avas 12alpha1, BD Horizon™
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 565970
Catalog No. BDB565970
Add to cart
Description
Description
FLK-1 Monoclonal antibody specifically detects FLK-1 in Mouse samples. It is validated for Flow cytometry.
Specifications
Specifications
FLK-1
Avas 12alpha1
Brilliant Violet 421
Purified
Rat
Rat WI, also known as Wistar (outbred) IgG2a, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions, and unconjugated antibody and free BD Horizon™ BV421 were removed.
Monoclonal
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Fetal liver kinase 1; CD309; Kdr; VEGF receptor-2; VEGFR-2
Mouse Flk-1 Recombinant Protein
25μg
Cell Biology
Primary
Murine
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.