Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FLNC Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP279816AF750
Description
FLNC Polyclonal specifically detects FLNC in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
FLNC | |
Polyclonal | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
ABP-280, ABP280A, ABP-280-like protein, ABPA, ABP-L, ABPLABP-L, gamma filamin, Actin-binding-like protein, filamin 2, filamin C, gamma, filamin C, gamma (actin binding protein 280), Filamin-2, filamin-C, FLJ10186, FLN2actin binding protein 280, FLNc, FLN-C, Gamma-filamin | |
Rabbit | |
Affinity Purified | |
RUO | |
Human, Mouse | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 750 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
FLNC | |
Recombinant protein made to an internal portion of the human FLNC protein (amino acids 270-468) [UniProt Q14315] | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction