Learn More
Research Products International CorpSupplier Diversity Partner PrepSafe Floating Micro-Tube Rack, Pink DFS Item
Manufacturer:  Research Products International CorpSupplier Diversity Partner 246726P
PrepSafe Racks protect samples from loss or contamination during incubation procedures or when chilled in an ice bucket. Unique locking tabs hold tubes securely in place and prevent lids from popping open during boiling applications. Racks are designed to float in a water bath or most 1000ml beakers. Manufactured from polypropylene and can be autoclaved. Each rack holds up to twelve 1.5-2.0ml micro-tubes.
Catalog No. 50-476-621