Learn More
Cole-Parmer Hedland H621-610 Flowmeter, spring-loaded in-line, for water, 1" NPT(M), 1 to 10 gpm  DFS Item
Manufacturer:  Cole-Parmer 0323145
These economical flowmeters are built with a spring-retained piston for direct, accurate readings regardless of the position you mount them. Use them to monitor hydraulic systems or chemical processes; the meters also work well to check pump and control valve performance. Note: Polysulfone is not suited for direct contact with sunlight.
Considering limit switches? Use to provide an output signal at a setpoint anywhere between 0 to 100% of full-scale. Use to control small pumps, valves, or alarms or to interface with a PLC.
Catalog No. 50-631-285