Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eagle™ Floor Model Corrosives Cabinet, One-Door Model

Store corrosives, up to 24 gallons, with an Eagle Corrosives Cabinet. This models sits securely on the floor and features one door for ease of use.

Supplier:  Eagle™ CRA1904X

 View more versions of this product

Catalog No. 19-120-714


Add to Cart

Alert:

Not recommended for use with sulfuric, nitric, or hydrochloric acid.


Description

Description

  • Suitable for storage of flammable and nonflammable acids and corrosive liquids
  • Rugged 18-gauge steel cabinets coated with tough blue epoxy powder on interior and exterior surfaces, including shelves, for extra protection against corrosive acids and caustics
  • Coated surfaces are superior to epoxy- or enamel-painted surfaces in case of spill or splash
  • Doors clearly labeled “CAUTION—CORROSIVES”
  • High-density polyethylene trays for shelves and bottom with drip edge contains liquid spills; removable for easy cleaning

Certifications

FM approved except models 17-120-2933, 19-120-714, and 19-120-715.

Compliance

Meet NFPA Code 30 and OSHA requirements.

Specifications

Specifications

4 gal. (15 L)
15 x 14.5 x 19.5 in. (38 x 36.8 x 49.53 cm)
57 cm
31.2 kg
45.7 cm
1
14.5 in.
22.5 in.
18 in.
NFPA, OSHA
17.5 in.
Blue
69 lb.
Storage Cabinet
49.53 cm
45 cm
38 cm
36.8 cm
19.5 in.
15 in.
Flammables
1
HDPE shelves and bottom trays with drip edge
Epoxy
Steel
Manual-close
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit