Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Florisil, 60-100 mesh, for column chromatography, ACROS Organics™

Click to view available options
Quantity:
250g
500g
1kg
5kg
Packaging:
Glass bottle
Plastic drum
1343-88-0
MgO3Si
100.39
MFCD00078249
ZADYMNAVLSWLEQ-UHFFFAOYSA-N
1343-88-0
MgO3Si
100.39
MFCD00078249
ZADYMNAVLSWLEQ-UHFFFAOYSA-N

Florisil, MgO3Si, CAS Number-1343-88-0, 1kg, White, 01,393; 09,223, 100.39, ZADYMNAVLSWLEQ-UHFFFAOYSA-N, 10, 5509, MFCD00078249, 60-100 mesh, for column chromatography, Glass bottle

Florisil, MgO3Si, CAS Number-1343-88-0, 1kg, White, 01,393; 09,223, 100.39, ZADYMNAVLSWLEQ-UHFFFAOYSA-N, 10, 5509, MFCD00078249, 60-100 mesh, for column chromatography, Glass bottle

for column chromatography
Melting Point >1500°C
Color White
Quantity 1kg
Packaging Glass bottle
Fieser 01,393; 09,223
Merck Index 10, 5509
Particle Size Particle size analysis (typical values): 60 mesh: 2.0%, 70 mesh: 20.0%, 80 mesh: 41.0%, 100 mesh: 30.0%, 140 mesh: 6.9%
Formula Weight 100.39
Physical Form Fine Powder
Chemical Name or Material Florisil™, 60-100 mesh, for column chromatography