Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Florisil™, Activated at 675°C, 60 to 100 Mesh, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2kg
Packaging:
Poly Pail
Wide Mouth Amber Glass Bottle
1343-88-0
MgO3Si
100.39
ZADYMNAVLSWLEQ-UHFFFAOYSA-N
1343-88-0
MgO3Si
100.39
ZADYMNAVLSWLEQ-UHFFFAOYSA-N

High quality chemicals for laboratory and specialized industrial use

    Suitable for Use in Chromatographic Cleanup of Pesticide Residues
Quantity 2kg
Mesh Size 60 to 100 Mesh
Packaging Poly Pail
Specific Gravity 1L = 2.51kg
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Florisil™, Activated at 675°C