Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flotillin-1 Rabbit anti-Human, Mouse, HRP, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP179022H

Catalog No. NBP179022H


Add to cart

Description

Description

Flotillin-1 Polyclonal antibody specifically detects Flotillin-1 in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

Flotillin-1
HRP
PBS with No Preservative
flotillin 1, flotillin-1, integral membrane component of caveolae
Rabbit
IgG
100 ul
Store at 4C in the dark.
Polyclonal
Human, Mouse
Western Blot
Western Blot
Affinity Purified
FLOT1
The immunogen recognized by this antibody maps to a region between residue 377 and 427 of human Flotillin 1 using the numbering given in entry NP_005794.1 (GeneID 10211).
Immunogen affinity purified
RUO
Primary
10211
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only